Orders LinkContactUsLinkComponent
Replacement caps
Replacement caps
Supplier:   QVF LABORTECHNIK
Replacement caps
Supplier:  QVF LABORTECHNIK
Certificates
Replacement caps
3 Options Available Below
Product Details & Documents
Pyrex®, phenoplast, black.
  • Excellent chemical resistance, excellent temperature resistance
  • Screw caps with PTFE-coated seal
  • Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • SVL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,90 €
    5,90 €
    1 * 1 ST
       5,90 €
  • SVL 20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,60 €
    6,60 €
    1 * 1 ST
       6,60 €
  • SVL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,00 €
    7,00 €
    1 * 1 ST
       7,00 €