Orders LinkContactUsLinkComponent
Test tubes
 :   DELTALAB
Test tubes
 :  DELTALAB
Test tubes
Test tubes
Polypropylene (PP), ultra clear.
  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable
Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.
Product Family Options
  • Capacity
    Length
    Ext.Ø
    Version
    Ø int.
  • 3 ml
    54,3 mm
    11,6 mm
    ungraduated
    9,9 mm
    1 * 2.000 ST
    1 * 2.000 ST - 129,00 €
    129,00 €
    1 * 2.000 ST
       129,00 €
    Specifications
    Cat. No.
    212-3320
  • 5 ml
    48,9 mm
    15,5 mm
    flat bottom, graduated
    13,7 mm
    1 * 5.000 ST
    1 * 5.000 ST - 366,00 €
    366,00 €
    1 * 5.000 ST
       366,00 €
    Specifications
    Cat. No.
    DELT401100
  • 5 ml
    74,2 mm
    11,8 mm
    graduated
    10,3 mm
    1 * 1.000 ST
    1 * 1.000 ST - 54,20 €
    54,20 €
    1 * 1.000 ST
       54,20 €
    Specifications
    Cat. No.
    212-3321
  • 5 ml
    74,2 mm
    12,9 mm
    ungraduated
    10,9 mm
    1 * 1.000 ST
    1 * 1.000 ST - 66,80 €
    66,80 €
    1 * 1.000 ST
       66,80 €
    Specifications
    Cat. No.
    DELT400700
  • 6 ml
    87,1 mm
    11,9 mm
    graduated
    10,3 mm
    1 * 1.000 ST
    1 * 1.000 ST - 73,60 €
    73,60 €
    1 * 1.000 ST
       73,60 €
    Specifications
    Cat. No.
    212-3322
  • 7 ml
    99,1 mm
    12,9 mm
    ungraduated
    10,9 mm
    1 * 500 ST
    1 * 500 ST - 57,10 €
    57,10 €
    1 * 500 ST
       57,10 €
    Specifications
    Cat. No.
    DELT401700
  • 10 ml
    95,3 mm
    15,4 mm
    graduated
    13,7 mm
    1 * 2.500 ST
    1 * 2.500 ST - 285,00 €
    285,00 €
    1 * 2.500 ST
       285,00 €
    Specifications
    Cat. No.
    DELT400900
  • 10 ml
    100 mm
    15,8 mm
    ungraduated
    13,7 mm
    1 * 500 ST
    1 * 500 ST - 63,30 €
    63,30 €
    1 * 500 ST
       63,30 €
    Specifications
    Cat. No.
    212-3326
Closures
  •  
    Colour
    Description
  • Natural
    Cap for 11 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 139,00 €
    139,00 €
    1 * 2.000 ST
       139,00 €
  •  
    Colour
    Description
  • Red
    Cap for 11 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 139,00 €
    139,00 €
    1 * 2.000 ST
       139,00 €
  •  
    Colour
    Description
  • Natural
    Cap for 15 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 161,00 €
    161,00 €
    1 * 2.500 ST
       161,00 €
  •  
    Colour
    Description
  • Green
    Cap for 12 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 117,00 €
    117,00 €
    1 * 2.000 ST
       117,00 €
  •  
    Colour
    Description
  • Green
    Cap for 16 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 143,00 €
    143,00 €
    1 * 2.500 ST
       143,00 €
  •  
    Colour
    Description
  • Natural
    Cap for 16 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 141,00 €
    141,00 €
    1 * 2.500 ST
       141,00 €
  •  
    Colour
    Description
  • Blue
    Cap for 12 and 13 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 80,90 €
    80,90 €
    1 * 2.000 ST
       80,90 €
  •  
    Colour
    Description
  • Green
    Cap for 12 and 13 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 80,90 €
    80,90 €
    1 * 2.000 ST
       80,90 €
  •  
    Colour
    Description
  • Natural
    Cap for 12 and 13 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 80,90 €
    80,90 €
    1 * 2.000 ST
       80,90 €
  •  
    Colour
    Description
  • Red
    Cap for 15 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 141,00 €
    141,00 €
    1 * 2.500 ST
       141,00 €
  •  
    Colour
    Description
  • Mauve
    Cap for 11 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 139,00 €
    139,00 €
    1 * 2.000 ST
       139,00 €
  •  
    Colour
    Description
  • Natural
    Cap for 15 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 141,00 €
    141,00 €
    1 * 2.500 ST
       141,00 €
  •  
    Colour
    Description
  • Natural
    Cap for 12 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 117,00 €
    117,00 €
    1 * 2.000 ST
       117,00 €
  •  
    Colour
    Description
  • Green
    Cap for 11 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 139,00 €
    139,00 €
    1 * 2.000 ST
       139,00 €
  •  
    Colour
    Description
  • Blue
    Cap for 12 mm tubes
    1 * 2.000 ST
    1 * 2.000 ST - 117,00 €
    117,00 €
    1 * 2.000 ST
       117,00 €
  •  
    Colour
    Description
  • Green
    Cap for 15 mm tubes
    1 * 2.500 ST
    1 * 2.500 ST - 141,00 €
    141,00 €
    1 * 2.500 ST
       141,00 €