Orders LinkContactUsLinkComponent
Test and sample tubes
 :   Anicrin
Test and sample tubes
 :  Anicrin
Test and sample tubes
Test and sample tubes
Clear PS or natural-coloured PP.
  • PS tubes: Non-autoclavable. Temperature resistant from −40 to +90 °C
  • PP tubes: Autoclavable up to 122 °C. Temperature resistant from −90 to +135 °C
  • Available with round or conical base
Precision moulded test tubes and sample tubes with round or conical base. Non sterile.
Product Family Options
  • Capacity
    Length
    Ext.Ø
    Material
    Type
  • 5 ml
    75 mm
    12 mm
    PS
    Round bottom
    1 * 5.000 ST
    1 * 5.000 ST - 210,00 €
    210,00 €
    1 * 5.000 ST
       210,00 €
  • 11 ml
    100 mm
    16 mm
    PS
    Round bottom
    1 * 2.000 ST
    1 * 2.000 ST - 168,00 €
    168,00 €
    1 * 2.000 ST
       168,00 €
  • 10 ml
    100 mm
    16 mm
    PS
    Conical bottom
    1 * 2.000 ST
    1 * 2.000 ST - 168,00 €
    168,00 €
    1 * 2.000 ST
       168,00 €
  • 11 ml
    100 mm
    16 mm
    PP
    Round bottom
    1 * 2.000 ST
    1 * 2.000 ST - 168,00 €
    168,00 €
    1 * 2.000 ST
       168,00 €