Orders LinkContactUsLinkComponent
Test tubes
Test tubes
Supplier:   KARTELL
Test tubes
Supplier:  KARTELL
Certificates
Test tubes
8 Options Available Below
Product Details & Documents
PS, transparent or PP, translucent.
  • PS: Good thermal and chemical resistance
  • PP: Excellent thermal and chemical resistance, autoclavable
  • Robust, economical
Universal tubes for a wide range of applications.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Material
    Type
    Availability
    Price
  • 3 ml
    55 mm
    11,5 mm
    PP
    Coagulometer
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 198,00 €
    198,00 €
    1 * 5.000 ST
       198,00 €
  • 5 ml
    70 mm
    11 mm
    PS
    RIA
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 45,10 €
    45,10 €
    1 * 1.000 ST
       45,10 €
  • 3 ml
    55 mm
    11,5 mm
    PS
    Coagulometer
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 44,50 €
    44,50 €
    1 * 1.000 ST
       44,50 €
  • 5 ml
    75 mm
    12 mm
    PS
    Cylindrical
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 48,70 €
    48,70 €
    1 * 1.000 ST
       48,70 €
  • 5 ml
    75 mm
    13 mm
    PS
    Cylindrical
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 245,00 €
    245,00 €
    1 * 4.000 ST
       245,00 €
  • 10 ml
    105 mm
    16 mm
    PS
    Conical
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 191,00 €
    191,00 €
    1 * 2.000 ST
       191,00 €
  • 10 ml
    100 mm
    16 mm
    PS
    Cylindrical
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 67,60 €
    67,60 €
    1 * 500 ST
       67,60 €
  • 20 ml
    150 mm
    16 mm
    PS
    Cylindrical
    Log in for availability
    Log in for availability
    1 * 850 ST
    1 * 850 ST - 190,00 €
    190,00 €
    1 * 850 ST
       190,00 €