Orders LinkContactUsLinkComponent
Test tubes
Test tubes
   Sustainable Options Available
Supplier:   Greiner Bio-One
Test tubes
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Test tubes
10 Options Available Below
Product Details & Documents
PS, clear or PP, transparent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
  • Matching push-in stoppers are optionally available
  • Can also be used as centrifuge tubes
Universal tubes for a wide range of laboratory and production applications.
Specifications for Product Family
Specifications
  • Catalog No:
  • 212-0361
  • GREI115201_2000
  • 212-3110
  • 391-3469
  • 212-3165
  • 212-3140
  • 212-3146
  • 212-3150
  • 212-0363
  • 212-3186
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
PP, with round bottom
Product Information
  • Capacity
    Length
    Ext.Ø
    Availability
    Price
  • 4 ml
    55 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 3.600 ST
    1 * 3.600 ST - 222,00 €
    222,00 €
    1 * 3.600 ST
       222,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0361
    Environmentally Preferable
    Environmental certification
  • 5 ml
    75 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 122,00 €
    122,00 €
    1 * 2.000 ST
       122,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI115201_2000
    Environmentally Preferable
    Environmental certification
  • 12 ml
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.600 ST
    1 * 1.600 ST - 175,00 €
    175,00 €
    1 * 1.600 ST
       175,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-3110
    Environmentally Preferable
    Environmental certification
  • 14 ml
    100 mm
    17 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 169,00 €
    169,00 €
    1 * 1.500 ST
       169,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    391-3469
    Environmentally Preferable
    Environmental certification
PS, with round bottom
Product Information
  • Capacity
    Length
    Ext.Ø
    Availability
    Price
  • 4 ml
    55 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 3.600 ST
    1 * 3.600 ST - 171,00 €
    171,00 €
    1 * 3.600 ST
       171,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-3165
    Environmentally Preferable
    Environmental certification
  • 5 ml
    75 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 169,00 €
    169,00 €
    1 * 2.000 ST
       169,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    212-3140
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 10 ml
    100 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 1.400 ST
    1 * 1.400 ST - 109,00 €
    109,00 €
    1 * 1.400 ST
       109,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-3146
    Environmentally Preferable
    Environmental certification
  • 12 ml
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.600 ST
    1 * 1.600 ST - 123,00 €
    123,00 €
    1 * 1.600 ST
       123,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-3150
    Environmentally Preferable
    Environmental certification
  • 14 ml
    100 mm
    17 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 95,30 €
    95,30 €
    1 * 1.500 ST
       95,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0363
    Environmentally Preferable
    Environmental certification
  • 20 ml
    152 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 188,00 €
    188,00 €
    1 * 1.500 ST
       188,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-3186
    Environmentally Preferable
    Environmental certification
Grip stoppers, PE
Product Information
  • Catalog #
    Colour
    Description
    For tubes
    Availability
    Price
  • Natural
    Grip stopper
    11 mm O.D.
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 17,60 €
    17,60 €
    1 * 500 ST
       17,60 €
Product Information
  • Catalog #
    Colour
    Description
    For tubes
    Availability
    Price
  • Natural
    Grip stopper
    14 mm O.D.
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 4,70 €
    4,70 €
    1 * 100 ST
       4,70 €
Product Information
  • Catalog #
    Colour
    Description
    For tubes
    Availability
    Price
  • Natural
    Grip stopper
    16 mm O.D.
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 4,80 €
    4,80 €
    1 * 100 ST
       4,80 €
Product Information
  • Catalog #
    Colour
    Description
    For tubes
    Availability
    Price
  • Natural
    Grip stopper
    12 mm O.D.
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 16,40 €
    16,40 €
    1 * 500 ST
       16,40 €