Culture tubes with screw cap, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, with DIN thread and red PBT screw caps with PTFE coated gaskets.
- Excellent chemical resistance
- High thermal resistance, sterilisation possible up to 1 bar overpressure at 121 °C
- Minimal thermal expansion giving relatively high resistance to temperature changes
Frequently Bought Together