ContactUsLinkComponent
Strips of 8 PCR tubes with attached cap strips
Strips of 8 PCR tubes with attached cap strips
Catalog # 211-3264
Supplier:  Brand
undefined
Strips of 8 PCR tubes with attached cap strips
Catalog # 211-3264
Supplier:  Brand
Specifications
  • Capacity:
    0,2 ml
  • Colour:
    Clear
  • Description:
    Strips of 8 PCR tubes with attached domed cap strips
  • Pk:
    125
Specifications
About this item
PP. Eight connected 0,2 ml tubes with attached domed cap strips.
  • Domed caps facilitate one-handed operation
  • Free from DNase, RNase and DNA
Colour: Clear
Colour: Clear