Order Entry
Austria
Orders LinkContactUsLinkComponent
Biotube™ system

Biotube™ system

Biotube™ system
Supplier:  Simport Scientific
Certificates
Biotube™ system
Biotube™ system

31 Options Available Below

Product Details & Documents

PP tube and rack system with tubes held in a standard 8×12 configuration.

  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Certified RNase, DNase, pyrogen- and DNA-free
  • Conforms to SBS standards
  • Caps are made of PE and are not autoclavable
  • Racks and tubes can be used over a wide temperature range, from −80 to +121 °C

These tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA.

Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as a self-standing support. It can also be placed in a refrigerator or freezer for improved air circulation around the tubes, or in water baths to allow controlled warming of the tubes contents.

Please note: Tubes have a gross volume of 1,2 ml but with a cap in place, they will hold 1,1 ml.

* PCR is a patented process of Hoffman-La Roche. Use of the PCR process requires a license.

Product Family Options

Tubes and caps

Product Information

  • Capacity
    Colour
    Availability
    Price
  • SIMPT100-28
    Individual plug caps, non sterile
    Individual plug caps, non sterile
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 410,00 €
    410,00 €
    1 * 4.800 ST
       410,00 €

  • 212-9551
    Individual tubes without writing surface, non sterile
    Individual tubes without writing surface, non sterile
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 56,10 €
    56,10 €
    1 * 960 ST
       56,10 €

  • SIMPT100-20LST
    Low surface tension individual tubes, non sterile
    Low surface tension individual tubes, non sterile
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 55,90 €
    55,90 €
    1 * 960 ST
       55,90 €

  • SIMPT100-30
    Plug caps, strips of 8, non sterile
    Plug caps, strips of 8, non sterile
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 134,00 €
    134,00 €
    1 * 600 ST
       134,00 €

  • SIMPT100-40
    Plug caps, strips of 12, non sterile
    Plug caps, strips of 12, non sterile
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 173,00 €
    173,00 €
    1 * 400 ST
       173,00 €

  • SIMPT100-35
    Strips of 8 plug caps, sterile
    Strips of 8 plug caps, sterile
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 178,00 €
    178,00 €
    1 * 600 ST
       178,00 €

  • 212-9552
    Strips of 8 tubes without writing surface
    Strips of 8 tubes without writing surface
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 56,40 €
    56,40 €
    1 * 120 ST
       56,40 €

  • SIMPT100-26
    Strips of 12 tubes, without writing surface, non sterile
    Strips of 12 tubes, without writing surface, non sterile
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 52,40 €
    52,40 €
    1 * 80 ST
       52,40 €

Racks and boxes

Product Information

  • Capacity
    Colour
    Availability
    Price
  • 720-0499
    Grid plate
    Grid plate
    1,2 ml
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,50 €
    59,50 €
    1 * 10 ST
       59,50 €

  • SIMPT100-60G
    Grid plate
    Grid plate
    1,2 ml
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,50 €
    59,50 €
    1 * 10 ST
       59,50 €

  • SIMPT100-60R
    Grid plate
    Grid plate
    1,2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,50 €
    59,50 €
    1 * 10 ST
       59,50 €

  • SIMPT100-60Y
    Grid plate
    Grid plate
    1,2 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,50 €
    59,50 €
    1 * 10 ST
       59,50 €

  • 212-9554
    Rack with 12 strips of 8 tubes
    Rack with 12 strips of 8 tubes
    1,2 ml
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 137,00 €
    137,00 €
    1 * 10 ST
       137,00 €

  • SIMPT100-3G
    Rack with 12 strips of 8 tubes
    Rack with 12 strips of 8 tubes
    1,2 ml
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 137,00 €
    137,00 €
    1 * 10 ST
       137,00 €

  • SIMPT100-3R
    Rack with 12 strips of 8 tubes
    Rack with 12 strips of 8 tubes
    1,2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 137,00 €
    137,00 €
    1 * 10 ST
       137,00 €

  • SIMPT100-3Y
    Rack with 12 strips of 8 tubes
    Rack with 12 strips of 8 tubes
    1,2 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 137,00 €
    137,00 €
    1 * 10 ST
       137,00 €

  • 212-9555
    Rack with 12 strips of 8 tubes, sterile
    Rack with 12 strips of 8 tubes, sterile
    1,2 ml
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 155,00 €
    155,00 €
    1 * 10 ST
       155,00 €

  • SIMPT100-4G
    Rack with 12 strips of 8 tubes, sterile
    Rack with 12 strips of 8 tubes, sterile
    1,2 ml
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 155,00 €
    155,00 €
    1 * 10 ST
       155,00 €

  • SIMPT100-4R
    Rack with 12 strips of 8 tubes, sterile
    Rack with 12 strips of 8 tubes, sterile
    1,2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 155,00 €
    155,00 €
    1 * 10 ST
       155,00 €

  • SIMPT100-4Y
    Rack with 12 strips of 8 tubes, sterile
    Rack with 12 strips of 8 tubes, sterile
    1,2 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 155,00 €
    155,00 €
    1 * 10 ST
       155,00 €

  • 212-9550
    Rack with 96 plain individual tubes
    Rack with 96 plain individual tubes
    1,2 ml
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 139,00 €
    139,00 €
    1 * 10 ST
       139,00 €

  • SIMPT100-1G
    Rack with 96 plain individual tubes
    Rack with 96 plain individual tubes
    1,2 ml
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 139,00 €
    139,00 €
    1 * 10 ST
       139,00 €

  • SIMPT100-1R
    Rack with 96 plain individual tubes
    Rack with 96 plain individual tubes
    1,2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 139,00 €
    139,00 €
    1 * 10 ST
       139,00 €

  • SIMPT100-1Y
    Rack with 96 plain individual tubes
    Rack with 96 plain individual tubes
    1,2 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 139,00 €
    139,00 €
    1 * 10 ST
       139,00 €

  • 212-9553
    Rack with 96 plain individual tubes, sterile
    Rack with 96 plain individual tubes, sterile
    1,2 ml
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 161,00 €
    161,00 €
    1 * 10 ST
       161,00 €

  • SIMPT100-2G
    Rack with 96 plain individual tubes, sterile
    Rack with 96 plain individual tubes, sterile
    1,2 ml
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 161,00 €
    161,00 €
    1 * 10 ST
       161,00 €

  • SIMPT100-2R
    Rack with 96 plain individual tubes, sterile
    Rack with 96 plain individual tubes, sterile
    1,2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 161,00 €
    161,00 €
    1 * 10 ST
       161,00 €

  • SIMPT100-2Y
    Rack with 96 plain individual tubes, sterile
    Rack with 96 plain individual tubes, sterile
    1,2 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 161,00 €
    161,00 €
    1 * 10 ST
       161,00 €

  • SIMPT100-50G
    Storage box only
    Storage box only
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 77,20 €
    77,20 €
    1 * 10 ST
       77,20 €

  • SIMPT100-50R
    Storage box only
    Storage box only
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 77,20 €
    77,20 €
    1 * 10 ST
       77,20 €

  • SIMPT100-50Y
    Storage box only
    Storage box only
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 77,20 €
    77,20 €
    1 * 10 ST
       77,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.