Microcentrifuge tubes with attached screw caps, 0,5/1,5/2,0 ml
Supplier: Brand
Certificates
About this item
PP, graduated, autoclavable, PP screw cap with silicone seal.
- Can withstand centrifugation to 17000×g
- Ideal for storage of serum and blood samples as well as for boiling and centrifugation
- Positive sealing screw cap with silicone seal
- Fast opening and closing with 1¹/₄ turn of cap
- Operating range: −196 to +121 °C, suitable for freezing samples in the gas phase of liquid nitrogen at −196 °C
*without graduations
Ordering information: Coloured cap inserts for identification are available separately.
Cap inserts, coloured
Product Information
Frequently Bought Together