ContactUsLinkComponent
Microcentrifuge tubes, 0,5 ml
Microcentrifuge tubes, 0,5 ml
Catalog # 211-2612
Supplier:  Brand
undefined
Microcentrifuge tubes, 0,5 ml
Catalog # 211-2612
Supplier:  Brand
Specifications
  • Type:
    with attached lid
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    0,5 ml
  • Colour:
    Colourless
  • Pk:
    1.000
  • RCF max.:
    10 000×g
Specifications
About this item
PP, autoclavable with attached cap.
  • Can withstand centrifugation up to 10000×g
  • Flat cap can be easily labelled and pierced
  • Tight fitting attached cap provides leak free seal, yet re-opens easily