Centrifuge tubes
Supplier: Brand
Certificates
About this item
AR® glass, conical bottom, beaded rim (except 525-0500 cone-pointed bottom, rimless).
- Withstands forces up to 3000×g
Frequently Bought Together
AR® glass, conical bottom, beaded rim (except 525-0500 cone-pointed bottom, rimless).