ContactUsLinkComponent
Laboratory trays
Catalog # 216-1466
Supplier:  KARTELL
undefined
Laboratory trays
Catalog # 216-1466
Supplier:  KARTELL
Specifications
  • Capacity:
    12 l
  • W×D×H:
    510×345×110 mm
  • Pk:
    1
Specifications
About this item
PP, translucent.
  • Excellent chemical and thermal resistance
  • Particularly deep design for increased stability
  • Versatile e.g. for transporting and storing laboratory utensils