Orders LinkContactUsLinkComponent
Laboratory trays
Laboratory trays
Supplier:   KARTELL
Laboratory trays
Supplier:  KARTELL
Certificates
Laboratory trays
Laboratory trays
8 Options Available Below
Product Details & Documents
PVC, white.
  • Thick walled and robust
  • Heat resistant up to 70 °C
  • Smooth surfaces allow easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Product Family Options
Product Information
  • W×D×H ext.
    W×D×H int.
    Availability
    Price
  • 245×195×50 mm
    200×150×45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,30 €
    27,30 €
    1 * 1 ST
       27,30 €
  • 305×250×60 mm
    250×200×60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • 385×325×75 mm
    320×260×70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,00 €
    57,00 €
    1 * 1 ST
       57,00 €
  • 420×375×85 mm
    350×300×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,70 €
    46,70 €
    1 * 1 ST
       46,70 €
  • 505×405×95 mm
    430×330×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,60 €
    95,60 €
    1 * 1 ST
       95,60 €
  • 600×495×100 mm
    520×420×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,00 €
    133,00 €
    1 * 1 ST
       133,00 €
  • 635×515×190 mm
    540×420×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 289,00 €
    289,00 €
    1 * 1 ST
       289,00 €
  • 765×620×110 mm
    675×530×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 218,00 €
    218,00 €
    1 * 1 ST
       218,00 €