Diagonal cutters
Supplier: KNIPEX WERKE
Jump to:
About this item
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
- High quality material and precise workmanship for long service life
- Precision cutting edges for soft and hard wire
- Cutting edges additionally induction hardened
: DIN ISO 5749. VDE-tested.
2 Options Available Below
Product Details & Documents
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
- High quality material and precise workmanship for long service life
- Precision cutting edges for soft and hard wire
- Cutting edges additionally induction hardened
: DIN ISO 5749. VDE-tested.
Product Family Options
Product Information
- AvailabilityPrice
- KNIP7005125Diagonal cutter, chrome plated, handles insulated with multi-component gripsDiagonal cutter, chrome plated, handles insulated with multi-component gripsLog in for availability1 * 1 ST33,70 €
- 238-5310Diagonal cutter, chrome plated, handles insulated with multi-component gripsDiagonal cutter, chrome plated, handles insulated with multi-component gripsLog in for availability1 * 1 ST40,00 €