Crucible tongs, stainless steel
Supplier: HAMMACHER KARL
Certificates
About this item
Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of crucibles made from a wide variety of materials.
- Double bend
- With grooved tips
- Acid-resistant
- Very good thermal resistance, autoclavable
Frequently Bought Together