Crucible tongs, nickel, straight
Catalog # 231-0353
Supplier: NICKEL ELECTRO
Specifications
- Length:200 mm
- Pk:1
Specifications
About this item
Pure nickel (99,6%). With grooved tips. For easy, safe handling of crucibles made from a wide variety of materials.
- Excellent corrosion resistance
- Good thermal and electrical conductivity
- Excellent thermal resistance