Order Entry
Austria
Orders LinkContactUsLinkComponent
Staining tile

Staining tile

8, Ø: 15 mm
Catalog # 631-9132
Supplier:   KARTELL
Staining tile
Catalog # 631-9132
Supplier:  KARTELL
Staining tile
Staining tile

8, Ø: 15 mm

Price
3,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

PS. This thin plate is suitable for staining applications.

W×D: 95×57 mm

Colour: clear

Specifications
  • No. of wells:
    8
  • Well size:
    Ø: 15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.