Orders LinkContactUsLinkComponent
Staining tiles
Staining tiles
Staining tiles
Certificates
Staining tiles
3 Options Available Below
Product Details & Documents
Glazed porcelain. These large tiles with cavities/wells are suitable for staining applications.
Colour: white.
Product Family Options
Product Information
  • No. of wells
    Well size
    W×D×H
    Verfügbarkeit
    Preis
  • 6
    Ø×H: 36×11 mm
    80×115×12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,60 €
    25,60 €
    1 * 1 ST
       25,60 €
  • 12
    Ø×H: 20×3 mm
    85×119×6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,80 €
    37,80 €
    1 * 1 ST
       37,80 €
  • 12
    Ø×H: 25×6 mm
    97×127×12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,90 €
    39,90 €
    1 * 1 ST
       39,90 €