Order Entry
Austria
ContactUsLinkComponent
Thermometer rack, SP Bel-Art
Thermometer rack, SP Bel-Art
Catalog # 620-0099
undefined
Thermometer rack, SP Bel-Art
Catalog # 620-0099

Specifications

  • Width:
    21.3 cm
  • Description:
    Thermometer rack, 25.1 cm H
  • Length:
    14.9 cm
  • Pk:
    1

Specifications

About this item

White PP. Stores up to 25 thermometers.

  • Supports thermometers vertically
  • Minimises any risk of breakage
  • Clearly organises your storage
  • Rapid access to the thermometers