Orders LinkContactUsLinkComponent
Temperature indicating strips
Supplier:   Testo
Temperature indicating strips
Supplier:  Testo
Certificates
Temperature indicating strips
6 Options Available Below
Product Details & Documents
Testoterm measuring points are self-adhesive, temperature-resistant films with heat-sensitive elements for monitoring a defined maximum temperature.
  • Irreversible change in colour within two seconds
  • Practical pack with 50 thermometer strips
  • One temperature level
W×D: 14×14 mm.
Specifications for Product Family
Specifications
  • Catalog No:
  • 620-9151
  • 620-9152
  • 620-9153
  • 620-9154
  • 620-9159
  • 620-9161
  • Storage temperature:
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Max. storage temperature +25 °C; Storage in refrigerator recommended
  • Temperature accuracy:
  • +43...+154 °C: ±1,5 °C
  • +43...+154 °C: ±1,5 °C
  • +43...+154 °C: ±1,5 °C
  • +43...+154 °C: ±1,5 °C
  • +43...+154 °C: ±1,5 °C
  • +43...+154 °C: ±1,5 °C
Specifications
Product Family Options
Product Information
  • Max. temp.
    W×D
    Availability
    Price
  • +65 °C
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9151
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C
  • +71 °C
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9152
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C
  • +77 °C
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9153
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C
  • +82 °C
    15×14 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9154
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C
  • +110 °C
    15×14 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9159
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C
  • +121 °C
    15×14 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 29,00 €
    29,00 €
    1 * 50 ST
       29,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    620-9161
    Storage temperature
    Max. storage temperature +25 °C; Storage in refrigerator recommended
    Temperature accuracy
    +43...+154 °C: ±1,5 °C