Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
26 Options Available Below
Product Details & Documents
This extensive range of low temperature thermometers is suitable for government verification as well as government calibration (620-1713). Toluene or pentane.
  • Total immersion
  • Colourless capillary - except 620-1047 (blue-reflecting capillary)
  • Three different fillings
Ø: 8 - 10 mm
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Model L33008DKD3 with DIN 12778.
Delivery information: Models 620-1047 and 620-0180 are government verified and supplied with a verification certificate.
Documents
Product Family Options
Division: 1,0 °C
Product Information
  • Range
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL33000-FL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −38...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 158,00 €
    158,00 €
    1 * 1 ST
       158,00 €
  • AMARL33000-FL-PTFE
    Low temperature laboratory thermometer, with PTFE coating
    Low temperature laboratory thermometer, with PTFE coating
    −38...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARL33000-FL/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −38...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARL33004-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,70 €
    59,70 €
    1 * 1 ST
       59,70 €
  • AMARL33004-TOL/DKD
    Low temperature laboratory thermometer, with DKD certificate
    Low temperature laboratory thermometer, with DKD certificate
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARL33004-TOL/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • AMARL33004-TOLDKD3
    Low temperature laboratory thermometer, with DKD3 certificate
    Low temperature laboratory thermometer, with DKD3 certificate
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 292,00 €
    292,00 €
    1 * 1 ST
       292,00 €
  • AMARL33008-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −100...+30 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,00 €
    75,00 €
    1 * 1 ST
       75,00 €
  • AMARL33008-TOL/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −100...+30 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 433,00 €
    433,00 €
    1 * 1 ST
       433,00 €
  • AMARL33008-TOLDKD3
    Low temperature laboratory thermometer, with DKD3 certificate
    Low temperature laboratory thermometer, with DKD3 certificate
    −100...+30 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 307,00 €
    307,00 €
    1 * 1 ST
       307,00 €
  • AMARL33008-TOLDKD5
    Low temperature laboratory thermometer, with DKD5 certificate
    Low temperature laboratory thermometer, with DKD5 certificate
    −100...+30 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 347,00 €
    347,00 €
    1 * 1 ST
       347,00 €
  • AMARL33018-PENT
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −200...+30 °C
    350 mm
    Pentane
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • AMARL33018-PENT/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −200...+30 °C
    350 mm
    Pentane
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 670,00 €
    670,00 €
    1 * 1 ST
       670,00 €
Division: 0.2 °C
Product Information
  • Range
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL33052-CB/DKD4
    Low temperature laboratory thermometer, with DKD certificate
    Low temperature laboratory thermometer, with DKD certificate
    −38...+50 °C
    400 mm
    Blue liquid
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARL33056-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −58...+30 °C
    400 mm
    Thallium
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 186,00 €
    186,00 €
    1 * 1 ST
       186,00 €
  • AMARL33058-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −50...+50 °C
    420 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • AMARL33058-TOL/CC3
    Low temperature laboratory thermometer, with calibration certificate
    Low temperature laboratory thermometer, with calibration certificate
    −50...+50 °C
    420 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 375,00 €
    375,00 €
    1 * 1 ST
       375,00 €
Division: 0.5 °C
Product Information
  • Range
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL33032-FL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −30...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • AMARL33032-FL-PTFE
    Low temperature laboratory thermometer, with PTFE coating
    Low temperature laboratory thermometer, with PTFE coating
    −30...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARL33032-FL/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −30...+50 °C
    220 mm
    Red liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 499,00 €
    499,00 €
    1 * 1 ST
       499,00 €
  • AMARL33038-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,70 €
    69,70 €
    1 * 1 ST
       69,70 €
  • AMARL33038-TOL/VC
    Low temperature laboratory thermometer, with verification certificate
    Low temperature laboratory thermometer, with verification certificate
    −50...+50 °C
    300 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 408,00 €
    408,00 €
    1 * 1 ST
       408,00 €
Division: 0,1 °C
Product Information
  • Range
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL33070-CB
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −30...+30 °C
    420 mm
    Blue liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • AMARL33074-CB
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −38...+50 °C
    550 mm
    Blue liquid
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 231,00 €
    231,00 €
    1 * 1 ST
       231,00 €
  • AMARL33076-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −58...+30 °C
    550 mm
    Thallium
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 227,00 €
    227,00 €
    1 * 1 ST
       227,00 €
  • AMARL33078-TOL
    Low temperature laboratory thermometer
    Low temperature laboratory thermometer
    −50...+50 °C
    580 mm
    Toluene
    8 - 10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 171,00 €
    171,00 €
    1 * 1 ST
       171,00 €