VWR®, Dry Block/Incubator Liquid-in-Glass Thermometers
Contains red organic liquid against white back glass, except for 620-0891 which has yellow back glass.
- For use in all standard heating blocks and incubators
- PTFE coating keeps liquid and glass encapsulated in the event of breakage
- Individually serialised
Accuracy: ±1,0 °C (620-0888: ±0,5 °C)
Frequently Bought Together