ContactUsLinkComponent
Nunc-Immuno™ stick
Nunc-Immuno™ stick
Catalog # 391-8283
   Low Manufacturing Impact
undefined
Nunc-Immuno™ stick
Catalog # 391-8283
   Low Manufacturing Impact
Specifications
  • Description:
    Nunc-Immuno™ stick with MaxiSorp™ paddle surface
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Pk:
    1.800
Specifications
About this item
PP tube and stick, PS paddle. Developed for ELISA and RIA tests in the field.
  • Gives reliable, semi-quantitative results with built-in negative control
  • Hydrophilic MaxiSorp™ paddle surface
  • PP tube for minimal adsorption of assay components
Paddle surface area: 5,2 cm²
External L×W (including lid): 49×12 mm