Orders LinkContactUsLinkComponent
Labelling tapes, set
Labelling tapes, set
Labelling tapes, set
Certificates
Labelling tapes, set
Labelling tapes, set
3 Options Available Below
Product Details & Documents
This all-purpose, self-adhesive tape is well suited for a wide variety of labelling applications. It can be written on with pen, pencil or permanent marker and is available in various colours, in three widths.
  • Temperature resistant from −45 to +121 °C
  • With removable adhesive
  • Oil, water and acid resistant
Set 1 contains two rolls each of white, yellow, blue, green, and red, and one roll of orange and pink.
Set 2 contains five rolls of white, four rolls of yellow, and three rolls each of blue, green, orange, red and pink.
Set 3 contains four rolls of white, and two rolls each of yellow, blue, green, orange, red and pink.
Length: 12,7 m
Documents
Product Family Options
Product Information
  • Colour
    Width
    Availability
    Price
  • 817-0026
    Tape set 1
    Tape set 1
    Assorted
    25,0 mm
    Log in for availability
    Log in for availability
    1 * 12 Rolle
    1 * 12 Rolle - 449,00 €
    449,00 €
    1 * 12 Rolle
       449,00 €
  • 817-0027
    Tape set 2
    Tape set 2
    Assorted
    13,0 mm
    Log in for availability
    Log in for availability
    1 * 24 Rolle
    1 * 24 Rolle - 532,00 €
    532,00 €
    1 * 24 Rolle
       532,00 €
  • 817-0044
    Tape set 3
    Tape set 3
    Assorted
    19,0 mm
    Log in for availability
    Log in for availability
    1 * 16 Rolle
    1 * 16 Rolle - 473,00 €
    473,00 €
    1 * 16 Rolle
       473,00 €