Order Entry
Austria
Orders LinkContactUsLinkComponent
Packaging tapes

Packaging tapes

Supplier:   3M
Packaging tapes
Supplier:  3M
Certificates
Packaging tapes

2 Options Available Below

Product Details & Documents

Universal adhesive tape for packaging, securing and labelling.

  • Excellent humidity resistance
  • Easy to tear during use
  • PE coating

Length: 50,0 m

Product Family Options

Product Information

  • Colour
    Width
    Availability
    Price
  • 819-1410
    Fabric adhesive tape
    Fabric adhesive tape
    White
    25,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 20,30 €
    20,30 €
    1 * 1 Rolle
       20,30 €

  • 819-1411
    Fabric adhesive tape
    Fabric adhesive tape
    Silver
    50,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 40,50 €
    40,50 €
    1 * 1 Rolle
       40,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.