Orders LinkContactUsLinkComponent
Sealing tape, PTFE
Supplier:   AMMERFLON
Sealing tape, PTFE
Supplier:  AMMERFLON
Certificates
Sealing tape, PTFE
Sealing tape, PTFE
3 Options Available Below
Product Details & Documents
This white tape is 0,1 mm thick and is used to seal threads. It is packed as a roll with protective cover for clean storage.
  • Heat-resistant from −150 to +260 °C
  • Excellent chemical resistance
  • Swell-resistant, allowing easy removal from the connection point even after continuous use
Product Family Options
Product Information
  • Width
    Availability
    Price
  • 291-1411
    Sealing tape, 12,0 m
    Sealing tape, 12,0 m
    12,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,90 €
    4,90 €
    1 * 1 ST
       4,90 €
  • 291-1412
    Sealing tape, 10,0 m
    Sealing tape, 10,0 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,50 €
    20,50 €
    1 * 1 ST
       20,50 €
  • AMMEPTFE-SEALTAPE
    Sealing tape, 10,0 m
    Sealing tape, 10,0 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 125 Rolle
    1 * 125 Rolle - 2 290,00 €
    2 290,00 €
    1 * 125 Rolle
       2 290,00 €