Tanks, cylindrical, lightweight
LLDPE, translucent with lid. Economical tanks with graduations. With protruding rim (flange). Wall thickness ~2,4 mm. Tank 216-0322 can also be used as an insert for 208 litre steel drums.
- Good chemical resistance
- Heat resistant from –100 to +120 °C
- Conform to the requirements of USP VI and FDA 21 CFR 177.1520
- Catalog No:
- 216-0322
- 216-0321
- 216-0317
- 216-0318
- 216-0320
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together