Orders LinkContactUsLinkComponent
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
Supplier:  BD Medical
Certificates
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
13 Options Available Below
Product Details & Documents
The BD Microlance™ 3 needles are designed for easy penetration, smooth gliding force during insertion/withdraw, and reduced cannula friction while sliding through tissue. These needles are available in a wide range of gauges, lengths, and bevel designs.
  • Recommended use: intradermal, subcutaneous, intramuscular, intravenous injections, and aspiration
  • BD Microlance™ needles can be connected to a luer slip adapter or luer-lock connector
  • Natural rubber latex is not part of the material formulation
  • Sterile, single-use design
Conventional needles with a regular size wall are less likely to flex than thin wall needles, due to the added cannula thickness. A thin wall needles inner diameter potentially allows use of a needle gauge that is one size smaller than that of a regular needle, while benefiting from the same flow rate.
Packaging: Individually packed in blister film and medical grade paper.
Product Family Options
Product Information
  • Gauge
    Length
    Ø
    Colour code
    Availability
    Price
  • 613-3832
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 27
    13 mm
    0,4 mm
    Grey
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 7,80 €
    7,80 €
    1 * 100 ST
       7,80 €
  • 613-3942
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 30
    13 mm
    0,3 mm
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8,60 €
    8,60 €
    1 * 100 ST
       8,60 €
  • BDAM304000UK
    BD Microlance™ 3 needle, thin wall, regular bevel
    BD Microlance™ 3 needle, thin wall, regular bevel
    G 30
    13 mm
    0,3 mm
    Yellow
    Discontinued
  • 613-0902
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 25
    16 mm
    0,5 mm
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,30 €
    9,30 €
    1 * 100 ST
       9,30 €
  • 615-0034
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 21
    16 mm
    0,8 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,70 €
    9,70 €
    1 * 100 ST
       9,70 €
  • 613-4952
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 25
    25 mm
    0,5 mm
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,30 €
    9,30 €
    1 * 100 ST
       9,30 €
  • 613-3924
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 23
    25 mm
    0,6 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8,90 €
    8,90 €
    1 * 100 ST
       8,90 €
  • 613-3935
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 21
    25 mm
    0,8 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,30 €
    9,30 €
    1 * 100 ST
       9,30 €
  • 613-3897
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 16
    40 mm
    1,6 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,60 €
    16,60 €
    1 * 100 ST
       16,60 €
  • 613-1688
    BD Microlance™ 3 needle, thin wall, regular bevel
    BD Microlance™ 3 needle, thin wall, regular bevel
    G 22
    40 mm
    0,7 mm
    Black
    Discontinued
  • BDAM303262
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 18
    40 mm
    Pink
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 10,20 €
    10,20 €
    1 * 100 ST
       10,20 €
  • BDAM303273
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 23
    40 mm
    0,64 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,70 €
    9,70 €
    1 * 100 ST
       9,70 €
  • 613-4970
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 21
    40 mm
    0,8 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,70 €
    9,70 €
    1 * 100 ST
       9,70 €
  • 613-3934
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 21
    50 mm
    0,8 mm
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 11,40 €
    11,40 €
    1 * 100 ST
       11,40 €
  • BDAM310205
    BD Microlance™ 3 needle, sterile
    BD Microlance™ 3 needle, sterile
    G 18
    50 mm
    1,20 mm
    Pink
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 19,80 €
    19,80 €
    1 * 100 ST
       19,80 €