VWR®, Transport Swabs with Medium
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
Collection and transport swabs for bacterial specimens with applicators and transport media.
- Bacteria will survive for 24 to 48 hours on the applicator and in the tube
- Tamper-proof seal, which turns white when open
- Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical wounds.
Certifications: Class IIa.
Packaging: Individually wrapped aluminium pouches under nitrogen ensuring longer shelf life.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Zugehörige Informationen
Frequently Bought Together