Orders LinkContactUsLinkComponent
Press-down stoppers
Supplier:   KARTELL
Press-down stoppers
Supplier:  KARTELL
Certificates
Press-down stoppers
Press-down stoppers
9 Options Available Below
Product Details & Documents
For sealing glass or plastic reagent and sample tubes.
  • PE, natural colour
  • Good chemical resistance
  • Economical
  • Convenient pack sizes
*conical
Product Family Options
Product Information
  • Colour
    Ø bottom
    Ø top
    Availability
    Price
  • Natural
    10,5 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 6,40 €
    6,40 €
    1 * 100 ST
       6,40 €
  • Natural
    14 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8,30 €
    8,30 €
    1 * 100 ST
       8,30 €
  • Natural
    14,5 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8,30 €
    8,30 €
    1 * 100 ST
       8,30 €
  • Natural
    16 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 9,20 €
    9,20 €
    1 * 100 ST
       9,20 €
  • Natural
    22 mm
    24 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 13,80 €
    13,80 €
    1 * 100 ST
       13,80 €
  • Natural
    28 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 18,90 €
    18,90 €
    1 * 100 ST
       18,90 €
  • Natural
    33 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 23,90 €
    23,90 €
    1 * 100 ST
       23,90 €
  • Natural
    38 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 25,30 €
    25,30 €
    1 * 100 ST
       25,30 €
  • Natural
    15,5 mm
    17 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 8,60 €
    8,60 €
    1 * 100 ST
       8,60 €