Silicone rubber stoppers, conical
Supplier: DEUTSCH NEUMANN
Jump to:
About this item
These stoppers are biologically inert and transparent.
- Resistant to temperatures from −60 to +220 °C and for short periods up to +250 °C
Certifications: DIN 12 871
42 Options Available Below
Product Details & Documents
These stoppers are biologically inert and transparent.
- Resistant to temperatures from −60 to +220 °C and for short periods up to +250 °C
Certifications: DIN 12 871
Documents
Product Family Options
With one hole
Product Information
- No. of HolesHeightHole ØØ bottomØ topAvailabilityPrice
With two holes
Product Information
- No. of HolesHeightHole ØØ bottomØ topAvailabilityPrice
Without holes
Product Information
- No. of HolesHeightHole ØØ bottomØ topAvailabilityPrice