Stoppers, rubber, with reversible edge
10,2 mm
Catalog # 217-0183
Supplier: Saint Gobain Life Sciences
Jump to:
Product Details & Documents
The white stoppers made of natural rubber guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers to be sealed are imperfect. They can be punctured and resealed after sampling, even after several uses.
- Outstanding sealing and protection from the outside atmosphere
- Suitable for puncturing
- Economical
Specifications
- Ø bottom:10,2 mm