Adapters with stopcock and connector
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With standard ground cone, standard ground stopcock with solid glass key and a straight or right angled tubing connector. The tubing connector has an external diameter of 8 mm.
Related Information
Adapter with cone NS 29/32
Technical Drawing
BSE TSE Glass Columns
BSE/TSE Certificates
Schott tubing
Datasheet
Frequently Bought Together