Orders LinkContactUsLinkComponent
Rods, glass
Rods, glass
Rods, glass
Certificates
Rods, glass
6 Options Available Below
Product Details & Documents
DURAN®, borosiicate glass.
  • Appropriately cut to 500 mm in length (cut ends)
  • Excellent chemical and thermal resistance
  • Permissible diameter tolerance of ±0.4 mm
Product Family Options
Product Information
  • Ext.Ø
    Length
    Availability
    Price
  • 3 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 37,70 €
    37,70 €
    1 * 10 ST
       37,70 €
  • 4 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 45,80 €
    45,80 €
    1 * 10 ST
       45,80 €
  • 5 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 58,30 €
    58,30 €
    1 * 10 ST
       58,30 €
  • 6 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 68,70 €
    68,70 €
    1 * 10 ST
       68,70 €
  • 7 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 70,60 €
    70,60 €
    1 * 10 ST
       70,60 €
  • 8 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 92,00 €
    92,00 €
    1 * 10 ST
       92,00 €