Order Entry
Austria
Orders LinkContactUsLinkComponent
Spatula

Spatula

Spatula, polypropylene, with two different shaped ends, 245 mm, 15 mm, White
Catalog # 441-0215
Supplier:   Brand
Spatula
Catalog # 441-0215
Supplier:  Brand
Spatula
Spatula

Spatula, polypropylene, with two different shaped ends, 245 mm, 15 mm, White

Price
19,50 €
Log in for availability
1 * 10 ST
Product Details & Documents

PP. Two differently shaped spatula ends (approx. 15 mm wide).

Specifications
  • Colour:
    White
  • Description:
    Spatula, polypropylene, with two different shaped ends
  • Length:
    245 mm
  • Blade width:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.