Order Entry
Austria
ContactUsLinkComponent
Magnetic stirring bar arrester, SP Bel-Art
Magnetic stirring bar arrester, SP Bel-Art
  442-0315
undefined
Magnetic stirring bar arrester, SP Bel-Art
  442-0315

 

  • Ø:
    51 mm
  • Width:
    9,5 mm
  • Pk:
    1

 

 

Polyethylene (9,5 mm) with radial magnet (Ø 51 mm). Position the arrester against the vessel bottom.

  • Prevents the stirring bar from being poured out with the liquid from a beaker or flask