Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, triangular | Avantor
VWR®, Magnetic stirring bars, triangular
VWR®, Magnetic stirring bars, triangular
Supplier:  VWR Collection
Certificates
VWR®, Magnetic stirring bars, triangular
VWR®, Magnetic stirring bars, triangular
9 Options Available Below
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the scraper-like action on the base of the container.
  • High turbulence, even at low speeds
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Product Family Options
Product Information
  • Length
    Ø
    Availability
    Price
  • 12 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,10 €
    24,10 €
    1 * 10 ST
       24,10 €
  • 20 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,70 €
    28,70 €
    1 * 10 ST
       28,70 €
  • 25 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 23,80 €
    23,80 €
    1 * 10 ST
       23,80 €
  • 35 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 38,00 €
    38,00 €
    1 * 10 ST
       38,00 €
  • 40 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 23,10 €
    23,10 €
    1 * 5 ST
       23,10 €
  • 50 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 26,00 €
    26,00 €
    1 * 5 ST
       26,00 €
  • 80 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 39,70 €
    39,70 €
    1 * 5 ST
       39,70 €
  • 110 mm
    36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,00 €
    50,00 €
    1 * 1 ST
       50,00 €
  • 136 mm
    36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,10 €
    62,10 €
    1 * 1 ST
       62,10 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.