VWR®, Magnetic Stirring Bars, Elliptical, Rare Earth
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
PTFE covered, rare earth magnetic core. The magnetic core is made from a combination of samarium and cobalt. The stirrers will continue to function at full strength for many years as the magnetism remains for longer. They are identified by an inert carbon black spot.
- Magnetic attraction is 1,5 to 2 times stronger than conventional stirrers
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Zugehörige Informationen
Frequently Bought Together