Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, rectangular
Supplier:  VWR Collection
Certificates
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular
4 Options Available Below
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Product Family Options
Product Information
  • Length
    Height
    Availability
    Price
  • 12 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 10,80 €
    10,80 €
    1 * 10 ST
       10,80 €
  • 25 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 19,40 €
    19,40 €
    1 * 10 ST
       19,40 €
  • 35 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 22,10 €
    22,10 €
    1 * 10 ST
       22,10 €
  • 50 mm
    7,5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 16,10 €
    16,10 €
    1 * 5 ST
       16,10 €