Distilling bridges
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Air cooled. Two vertical cones and a ground socket for a distillation thermometer with 55 mm immersion length.
Frequently Bought Together