Express Eingabe
Austria
ContactUsLinkComponent
Reinraumpapier, TexWrite® 22
Reinraumpapier, TexWrite® 22
Catalog # 818-0132
Supplier:  Texwipe
undefined
Reinraumpapier, TexWrite® 22
Catalog # 818-0132
Supplier:  Texwipe
Specifications
  • Farbe:
    Blau
  • Bezeichnung:
    Papier, A4
  • Seq:
    01
  • Modell:
    TX5811
  • VE:
    2.500
Specifications
About this item
TexWrite® 22, latex-saturated cellulose cleanroom documentation sheets without organic fillers.
  • Ideal for note taking, equipment manuals, and work instructions
  • Can be used with laser printers and photocopiers
Format: A4 Reinraumverbundpapier.
Packaging: 250 Blatt pro Packung, 10 Packungen pro Karton (Bag-Within-A-Bag®).