Orders LinkContactUsLinkComponent
Nalgene™ Staining Boxes
Nalgene™ Staining Boxes
   Sustainable Options Available
Nalgene™ Staining Boxes
   Sustainable Options Available
Certificates
Nalgene™ Staining Boxes
2 Options Available Below
Product Details & Documents
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a –70 to +70 °C (–94 to +158 °F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
Thermo Scientific™ Nalgene™ staining box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 189×189 mm (4,8×4,8") gel along with 500 ml of liquid.
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-8271
  • NALG715705-0001
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Material
    Capacity
    W×D×H
    Availability
    Price
  • 216-8271
    Staining boxes
    Staining boxes
    PMP
    500 ml
    127×127×508 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 102,00 €
    102,00 €
    1 * 2 ST
       102,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8271
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • NALG715705-0001
    Replacement plug for 5705-1010 and 2020
    Replacement plug for 5705-1010 and 2020
    PMP
    — mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,60 €
    32,60 €
    1 * 1 ST
       32,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG715705-0001
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact