Orders LinkContactUsLinkComponent
Pharmacist spoons
Pharmacist spoons
Supplier:   BOCHEM
Pharmacist spoons
Supplier:  BOCHEM
Certificates
Pharmacist spoons
Pharmacist spoons
4 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoons for a wide range of uses.
  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Spoon size
    Availability
    Price
  • BOCH3360
    Spoon in scoop shape
    Spoon in scoop shape
    170 mm
    40×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €
  • 231-1281
    Spoon with hole
    Spoon with hole
    200 mm
    30×26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,30 €
    5,30 €
    1 * 1 ST
       5,30 €
  • 231-1282
    Standard spoon
    Standard spoon
    150 mm
    30×15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,80 €
    2,80 €
    1 * 1 ST
       2,80 €
  • BOCH3385
    Standard spoon
    Standard spoon
    150 mm
    20×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €