Orders LinkContactUsLinkComponent
Measuring spoons, set
Measuring spoons, set
Set of 4 measuring spoons
Catalog # 231-1320
Supplier:   BOCHEM
Measuring spoons, set
Catalog # 231-1320
Supplier:  BOCHEM
Measuring spoons, set
Measuring spoons, set
Set of 4 measuring spoons
Price
11,40 €
Log in for availability
1 * 1 SET
Product Details & Documents
Stainless steel 18/10. Practical and convenient set of four measuring spoons.
  • Heat and chemical resistant
  • Ringed set keeps all sizes together
Delivery information: Each set includes 1,25 ml, 2,5 ml, 5 ml and 15 ml spoons.
Specifications
  • Description:
    Set of 4 measuring spoons