Orders LinkContactUsLinkComponent
Laboratory spoons
Laboratory spoons
Supplier:   BOCHEM
Laboratory spoons
Supplier:  BOCHEM
Certificates
Laboratory spoons
Laboratory spoons
5 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design
Product Family Options
Product Information
  • Length
    Spoon size
    Availability
    Price
  • 180 mm
    55×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,50 €
    4,50 €
    1 * 1 ST
       4,50 €
  • 195 mm
    60×45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,70 €
    4,70 €
    1 * 1 ST
       4,70 €
  • 280 mm
    85×70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • 120 mm
    40×26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €
  • 135 mm
    45×31 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,70 €
    2,70 €
    1 * 1 ST
       2,70 €