Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spoons
Weighing spoons with spatula end
Weighing spoons with spatula end
Weighing spoons with spatula end
Catalog # 231-0273
Supplier:
HAMMACHER KARL
Expand image
Weighing spoons with spatula end
Catalog # 231-0273
Supplier:
HAMMACHER KARL
Specifications
Length:
150 mm
Pk:
1
Spoon size:
26×16 mm
Specifications
About this item
Product Details
WIRONIT steel, anti-magnetic.
Product Details