Order Entry
Austria
ContactUsLinkComponent
Mixing spatula, spoon end/sharp end
Mixing spatula, spoon end/sharp end
Catalog # 231-0253
Supplier:  HAMMACHER KARL
undefined
Mixing spatula, spoon end/sharp end
Catalog # 231-0253
Supplier:  HAMMACHER KARL

Specifications

  • Description:
    No. 5
  • Length:
    190 mm
  • Blade width:
    10 mm
  • Pk:
    1
  • Blade length:
    20 mm

Specifications

About this item

Mixing spatula No. 5, made of WIRONIT steel.