Orders LinkContactUsLinkComponent
Cement spatulas
 :   BOCHEM
Cement spatulas
 :  BOCHEM
Cement spatulas
Cement spatulas
Stainless steel. Spatulas with single or double end.
Double spatula features one tappered end.
Product Family Options
  • Description
    Length
    Blade length
    Blade width
  • Double spatula
    150 mm
    35 mm
    6 mm
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €
  • Single spatula with tapered end
    150 mm
    35 mm
    6 mm
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €
  • Single spatula with straight end
    150 mm
    35 mm
    6 mm
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €