Orders LinkContactUsLinkComponent
Double spatulas, micro
Supplier:   RSG
Double spatulas, micro
Supplier:  RSG
Certificates
Double spatulas, micro
Double spatulas, micro
21 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 100 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 35,90 €
    35,90 €
    1 * 5 ST
       35,90 €
  • 130 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,50 €
    36,50 €
    1 * 5 ST
       36,50 €
  • 150 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 37,00 €
    37,00 €
    1 * 5 ST
       37,00 €
  • 185 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 37,60 €
    37,60 €
    1 * 5 ST
       37,60 €
  • 210 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 38,20 €
    38,20 €
    1 * 5 ST
       38,20 €
  • 150 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 150 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 185 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 150 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 185 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 210 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,60 €
    7,60 €
    1 * 1 ST
       7,60 €
  • 130 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,70 €
    36,70 €
    1 * 5 ST
       36,70 €
  • 100 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,50 €
    36,50 €
    1 * 5 ST
       36,50 €
  • 185 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,80 €
    36,80 €
    1 * 5 ST
       36,80 €
  • 150 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 20,00 €
    20,00 €
    1 * 5 ST
       20,00 €
  • 130 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 19,00 €
    19,00 €
    1 * 5 ST
       19,00 €
  • 100 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 18,10 €
    18,10 €
    1 * 5 ST
       18,10 €
  • 100 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 35,90 €
    35,90 €
    1 * 5 ST
       35,90 €
  • 130 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,50 €
    36,50 €
    1 * 5 ST
       36,50 €
  • 210 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 37,60 €
    37,60 €
    1 * 5 ST
       37,60 €
  • 130 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,50 €
    36,50 €
    1 * 5 ST
       36,50 €