Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
Supplier:   RSG
Spoon spatulas, micro
Supplier:  RSG
Certificates
Spoon spatulas, micro
Spoon spatulas, micro
4 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • With round handle and one bent spoon
  • Excellent temperature resistance, autoclavable
  • Good resistance to chemicals
  • Very flexible
Documents
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 130 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,00 €
    4,00 €
    1 * 1 ST
       4,00 €
  • 150 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 20,20 €
    20,20 €
    1 * 5 ST
       20,20 €
  • 185 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €
  • 210 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,40 €
    4,40 €
    1 * 1 ST
       4,40 €